Learn More
AdipoGen Sunitinib . malate (500 mg)
SDP
Catalog No. 502054083
Encompass_Preferred
Chemical. CAS: 341031-54-7. Formula: C22H27FN4O2 . C4H6O5. MW: 398.5 . 134.1. Potent ATP-competitive and cell permeable multi-targeted receptor tyrosine kinase (RTK) inhibitor targeting VEGFR and PDGFR-beta. Inhibits FLK1 (Ki=9nM), PDGFRbeta (Ki=8nM) and FLT3. It is at least 10-fold selective for FLK1 and PDGFRbeta over a variety of tyrosine kinases in a panel, including EGFR, Cdk2, Met, IGFR-1, Abl and Src. Inhibits the cellular receptor phosphorylation of FLT3, RET and CSF-1R. Also shown to inhibit c-Kit. Exhibits potent antiangiogenic and antitumor activity in multiple xenograft models.
Catalog No. 50-205-4083 Supplier AdipoGen Supplier No. AGCR13707M500
Chemical. CAS: 341031-54-7. Formula: C22H27FN4O2 . C4H6O5. MW: 398.5 . 134.1. Potent ATP-competitive and cell permeable multi-targeted receptor tyrosine kinase (RTK) inhibitor targeting VEGFR and PDGFR-beta. Inhibits FLK1 (Ki=9nM), PDGFRbeta (Ki=8nM) and FLT3. It is at least 10-fold selective for FLK1 and PDGFRbeta over a variety of tyrosine kinases in a panel, including EGFR, Cdk2, Met, IGFR-1, Abl and Src. Inhibits the cellular receptor phosphorylation of FLT3, RET and CSF-1R. Also shown to inhibit c-Kit. Exhibits potent antiangiogenic and antitumor activity in multiple xenograft models.