Learn More
MSC Sunnex Swivel Caster, 50 Lb Capacity, Nylon Wheel, Threaded Stem Mount
Catalog No. 5000536669
Encompass
This Sunnex swivel caster is designed for equipment requiring mobility and leveling capabilities. Featuring a 50 lb capacity and a durable nylon wheel, it offers smooth 360-degree motion and anti-vibration control. The threaded stem mount allows for easy installation and precise hand leveling adjustment, making it suitable for various industrial and manufacturing applications.
  • High quality nylon wheel
  • Dual mounting designs
  • 360 degree freedom of motion
  • Anti-vibration control with NBR pad
  • Easy hand leveling adjustment
Catalog No. 50-005-36669 Supplier MSC Supplier No. 86022852
This Sunnex swivel caster is designed for equipment requiring mobility and leveling capabilities. Featuring a 50 lb capacity and a durable nylon wheel, it offers smooth 360-degree motion and anti-vibration control. The threaded stem mount allows for easy installation and precise hand leveling adjustment, making it suitable for various industrial and manufacturing applications.
  • High quality nylon wheel
  • Dual mounting designs
  • 360 degree freedom of motion
  • Anti-vibration control with NBR pad
  • Easy hand leveling adjustment