Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sunset Yellow FCF, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013804 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3804 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3804 Supplier MilliporeSigma Supelco Supplier No. 6877525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sunset Yellow FCF azo-dye is generally used as a colorant in drug industries and in common foods.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2783-94-0
Physical Form Neat
Linear Formula C16H10N2 Na2O7S2
MDL Number MFCD00036437
Melting Point >300°C
Synonym Food Yellow 3; Orange Yellow S
Type Analytical Standard
Quantity 25 mg
Formula Weight 452.37
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.