Learn More
Nacalai USA Inc SunShell C18, 2.6 um (2.1 mmID x 75 mm)
Supplier:  Nacalai USA Inc CB6951
Encompass
SunShell HPLC columns are manufactured using innovative core shell particle technology that provides significantly increased column efficiency and chromatographic resolution without the high back pressure seen in sub-2 um columns.
Catalog No. NC1835038