Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich SunTronic™ silver nanoparticle ink for inkjet printing

Catalog No. 9019712ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-197-12ML 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-197-12ML Supplier Sigma Aldrich Supplier No. 9019712ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

SunTronic EMD5730 is formulated for inkjet printing of conductive traces onto a variety of substrates. EMD5730 is a solvent-based silver nanoparticle ink which demonstrates superior conductivity, stable jetting performance and adhesion to ITO coated glass.EMD5730 can be used in the manufacture of touch panel displays, printed RFID antennae, smart cards, sensors, security labels, touch switches and other printed electronics applications.SunTronic(R) silver nanoparticle ink for inkjet printing (SunTronic(R) EMD5730) is a 3D additive printing technology that is used in a variety of applications such as flexible microsystems, and electronics. It is printed on a liquid crystal polymeric substrate for the formation of a low cost, adaptable tooling for effective deposition of a wide range of materials.SunTronic is a registered trademark of Sun Chemical, Inc.

Specifications

Molecular Formula Ag
Linear Formula Ag
Quantity 2 mL
Synonym Ag ink; Conductive ink; Silver ink; SunTronic™EMD5730
Molecular Weight (g/mol) 107.87
Biological Activity Kidney
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.