Learn More
Medchemexpress LLC HY-132842 5mg , Sunvozertinib CAS:2370013-12-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1328425MG
Encompass_Preferred
HY-132842 5mg Sunvozertinib CAS: 2370013-12-8 Sunvozertinib (DZD9008) is a potent ErbBs (EGFR, Her2, especially mutant forms) and BTK inhibitor. Sunvozertinib shows IC50s of 20.4, 20.4, 1.1, 7.5, and 80.4 nM for EGFR exon 20 NPH insertion, EGFR exon 20 ASV insertion, EGFR L858R and T790M mutations, and Her2 Exon20 YVMA, and EGFR WT A431, respectively (patent WO2019149164A1, example 52). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1379