Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SuO-Glu-Val-Cit-PAB-MMAE | 1895916-24-1 | 95.0% | 1334.60 | C67H103N11O17 | 10 MG
SDP

Catalog No. 5000202029
Encompass_Preferred

SuO-Glu-Val-Cit-PAB-MMAE is a drug-linker conjugate combining a cleavable Glu-Val-Cit-PAB linker with the tubulin inhibitor MMAE for use in antibody-drug conjugate (ADC) synthesis. The product is supplied as a solid for research use only and requires low-temperature storage and protected handling.

  • Cleavable Glu-Val-Cit-PAB linker enabling enzymatic payload release.
  • Contains potent tubulin inhibitor payload (MMAE) for ADC development.
  • High purity (≈95.0%) suitable for conjugation and analytical work.
  • Soluble in DMSO; stock solution preparation guidance provided.
  • Store at -80°C and protect from light; unstable in solution, freshly prepare solutions before use.

Catalog No. 50-002-02029 Supplier Medchemexpress LLC Supplier No. HY13098910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SuO-Glu-Val-Cit-PAB-MMAE is a drug-linker conjugate combining a cleavable Glu-Val-Cit-PAB linker with the tubulin inhibitor MMAE for use in antibody-drug conjugate (ADC) synthesis. The product is supplied as a solid for research use only and requires low-temperature storage and protected handling.

  • Cleavable Glu-Val-Cit-PAB linker enabling enzymatic payload release.
  • Contains potent tubulin inhibitor payload (MMAE) for ADC development.
  • High purity (≈95.0%) suitable for conjugation and analytical work.
  • Soluble in DMSO; stock solution preparation guidance provided.
  • Store at -80°C and protect from light; unstable in solution, freshly prepare solutions before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.