Learn More
Medchemexpress LLC SuO-Val-Cit-PAB-MMAE | 99.2% | 1264.51 g/mol | C63H97N11O16 | 100 MG
SDP
Catalog No. 5000212375
Encompass_Preferred
SuO-Val-Cit-PAB-MMAE is a drug-linker conjugate for antibody-drug conjugate research that links the antimitotic payload monomethyl auristatin E (MMAE) to a protease-cleavable SuO-Val-Cit-PAB peptide linker, enabling intracellular release of the cytotoxic payload. The compound is supplied as a solid reagent for use as a building block in ADC synthesis and preclinical studies.
  • Protease-cleavable SuO-Val-Cit-PAB linker for targeted payload release.
  • Contains monomethyl auristatin E (MMAE) as the cytotoxic payload.
  • High purity suitable for research applications.
  • DMSO soluble; dissolve with ultrasonic and warming as needed.
  • Stored under inert atmosphere at -80°C to preserve stability.
  • Supplied in multiple package sizes for research flexibility.
Catalog No. 50-002-12375 Supplier Medchemexpress LLC Supplier No. HY100566100MG
SuO-Val-Cit-PAB-MMAE is a drug-linker conjugate for antibody-drug conjugate research that links the antimitotic payload monomethyl auristatin E (MMAE) to a protease-cleavable SuO-Val-Cit-PAB peptide linker, enabling intracellular release of the cytotoxic payload. The compound is supplied as a solid reagent for use as a building block in ADC synthesis and preclinical studies.
  • Protease-cleavable SuO-Val-Cit-PAB linker for targeted payload release.
  • Contains monomethyl auristatin E (MMAE) as the cytotoxic payload.
  • High purity suitable for research applications.
  • DMSO soluble; dissolve with ultrasonic and warming as needed.
  • Stored under inert atmosphere at -80°C to preserve stability.
  • Supplied in multiple package sizes for research flexibility.