Learn More
STRATASYS INC PACK OF 1 SUP706 B, SUPPORT , 4 KG
Catalog No. NC2816984
Encompass
SUP706B gives the freedom to design intricate and delicate multi-material parts that can be easily cleaned. The automated Support removal process enables you to clean internal cavities and delicate features that water-jetting might miss or damage. It greatly reduces labor time and increases productivity.
Catalog No. NC2816984 Supplier STRATASYS INC Supplier No. OBJ18016
SUP706B gives the freedom to design intricate and delicate multi-material parts that can be easily cleaned. The automated Support removal process enables you to clean internal cavities and delicate features that water-jetting might miss or damage. It greatly reduces labor time and increases productivity.