Learn More
Cellpath Supa Mega Slim Tissue Processing Cassettes
SDP
Catalog No. 22222131
Change view
Click to view available options
Color:
Green
Catalog No. Color
22-222-131 Green
1 options
This item is not returnable. View return policy
Catalog No. 22-222-131 Supplier Cellpath Supplier No. EAN010402A
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Supa Mega Slim is being widely adopted in clinical, research and toxicology laboratories. The cassette is similar to the Supa Mega Mothership in that its internal chamber is one-third of the depth of the conventional Supa Mega Cassette. Through using the Slim, casting can be done in only 20 minutes at -8°C.
  • Supa Mega Slim Cassette is being widely adopted in clinical, research and toxicology laboratories
  • Internal chamber is one-third of the depth of the conventional Supa Mega Cassette
  • This thinner depth is recommended in accordance with recommendations of HER2 testing in the UK and ASCO/CAP in the USA2,3, through reducing specimen thickness and shortening the processing time
  • Better reagent flow and improved turnaround times
  • Casting can be done in 20 minutes at -8°C
  • Less wax used during the embedding process
  • Large colour range available
  • Specific base moulds
Specifications
Color Green