Learn More
Cellpath Supa Mega Tissue Processing Cassettes
SDP
Catalog No. p-4908708
Change view
Click to view available options
Color:
Blue
Gray
Green
Peach
Pink
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-222-016 Green
22-222-020 Pink
22-222-017 Gray
22-222-015 White
22-222-021 Yellow
22-222-018 Blue
22-222-022 Red
22-222-023 Peach
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-222-016 Supplier Cellpath Supplier No. EAG010402A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Widely adopted in research and toxicology laboratories. CellPath Supa Mega Tissue Processing Cassettes are ideal for embedding and processing larger tissue samples of prostrate, brain, breast and eye.
  • Plastic hinged lid
  • Rigid, durable
  • Excellent writing surfaces
  • Internal chamber is three times depth of the Supa Mega Slim and Supa Mega Mother Ship
  • Available in a range of colors
Specifications
Quantity 100/Pk.
Color Green