Learn More
Electron Microscopy Sciences Supa Mega Cassette, Blue
SDP
Catalog No. 5033289
Encompass_Preferred
Extremely useful for research and toxicology laboratories. Ideal for prostate, brain, breast tissue and eyes. Processing cassettes ideal for prostate, brain, breast tissue and eyes.
Color: Blue
Catalog No. 50-332-89 Supplier Electron Microscopy Sciences Supplier No. 70065B
Extremely useful for research and toxicology laboratories. Ideal for prostate, brain, breast tissue and eyes. Processing cassettes ideal for prostate, brain, breast tissue and eyes.
Color: Blue