Learn More
Electron Microscopy Sciences Supa Mega Cassette, Green
SDP
Catalog No. 5033288
Encompass_Preferred
Extremely useful for research and toxicology laboratories. Ideal for prostate, brain, breast tissue and eyes. Processing cassettes ideal for prostate, brain, breast tissue and eyes.
Color: Green
Catalog No. 50-332-88 Supplier Electron Microscopy Sciences Supplier No. 70065G
Extremely useful for research and toxicology laboratories. Ideal for prostate, brain, breast tissue and eyes. Processing cassettes ideal for prostate, brain, breast tissue and eyes.
Color: Green