Learn More
Electron Microscopy Sciences Supa Mega Cassette, Pink
SDP
Catalog No. 5033287
Encompass_Preferred
Extremely useful for research and toxicology laboratories. Ideal for prostate, brain, breast tissue and eyes. Processing cassettes ideal for prostate, brain, breast tissue and eyes.
Color: Pink
Catalog No. 50-332-87 Supplier Electron Microscopy Sciences Supplier No. 70065P
Extremely useful for research and toxicology laboratories. Ideal for prostate, brain, breast tissue and eyes. Processing cassettes ideal for prostate, brain, breast tissue and eyes.
Color: Pink