Learn More
Electron Microscopy Sciences Supa Mega Cassette, Yellow
SDP
Catalog No. 5033291
Encompass_Preferred
Extremely useful for research and toxicology laboratories. Ideal for prostate, brain, breast tissue and eyes. Processing cassettes ideal for prostate, brain, breast tissue and eyes.
Color: Yellow
Catalog No. 50-332-91 Supplier Electron Microscopy Sciences Supplier No. 70065Y
Extremely useful for research and toxicology laboratories. Ideal for prostate, brain, breast tissue and eyes. Processing cassettes ideal for prostate, brain, breast tissue and eyes.
Color: Yellow