Learn More
Electron Microscopy Sciences SupaMega Stainless Steel Base Molds 36x36x10 mm, 10 mm height
SDP
Catalog No. 5033311
Encompass_Preferred
The SupaMega range of stainless steel base molds have extra deep side walls allowing wax to be decanted into the cassettes during embedding without the risk of the usual leakage of wax. The smaller faced mold also saves wax on medium sized blocks too large to fit into standard cassettes negating the use of full size molds.
Size: 36 x 36 mm
Height: 10 mm
Catalog No. 50-333-11 Supplier Electron Microscopy Sciences Supplier No. 6235436
The SupaMega range of stainless steel base molds have extra deep side walls allowing wax to be decanted into the cassettes during embedding without the risk of the usual leakage of wax. The smaller faced mold also saves wax on medium sized blocks too large to fit into standard cassettes negating the use of full size molds.
Size: 36 x 36 mm
Height: 10 mm