Learn More
ATCC SUP-B15; Lymphoblastic Leukemia; Human (Homo sapiens)
Catalog No. 502383784
Encompass
SUP-B15 is a B lymphoblast cell line that was isolated from the marrow of a White, 8-year-old, male patient with acute lymphoblastic leukemia (ALL). This cell line was deposited by SD Smith. It can be used in immunology and immune system disorder research.
Catalog No. 50-238-3784 Supplier ATCC Supplier No. CRL1929
SUP-B15 is a B lymphoblast cell line that was isolated from the marrow of a White, 8-year-old, male patient with acute lymphoblastic leukemia (ALL). This cell line was deposited by SD Smith. It can be used in immunology and immune system disorder research.