Learn More
Johnstone Supply SUPCO CHART PAPER (C)
Catalog No. NC9088441
Encompass_Preferred
Supco CR87-17 Temperature Recording Chart Paper; •7-Day Chart •Operating Range: -30°C to 10°C •Quantity: 60 Charts
Catalog No. NC9088441 Supplier Johnstone Supply Supplier No. CR8717
Supco CR87-17 Temperature Recording Chart Paper; •7-Day Chart •Operating Range: -30°C to 10°C •Quantity: 60 Charts