Learn More
MilliporeSigma™ Supelco™ Supel™ BioSPME 96-Pin Device
Catalog No. 01258420 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Pk.
10 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-420 10 Pk.
01-258-419 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-420 Supplier MilliporeSigma™ Supelco™ Supplier No. 59683U
Add to Cart
Add to Cart
The Supel™ BioSPME 96-Pin Device combines BioSPME technology with high throughput free fraction determination for plasma protein binding studies and other free analyte analyses commonly performed by pharma and clinical laboratories.
This fast, micro-sample prep device comprises a 96-pin universal format that fits into the wells of standard 96-well plates. The pins are coated with C18 particles and a patented binder that allows for small analytes of interest to bind, while larger macromolecules cannot. This extraction technique is shown to be more than three times faster than rapid equilibrium dialysis methods with improved data quality. Supel™ BioSPME 96-Pin Device workflow can be performed on automation robots via handling with standard gripper paddles, or alternatively can be achieved using a manual technique.
TRUSTED_SUSTAINABILITY
Specifications
Type SPME Pin Device
For Use With (Application) Clinical, Pharmaceutical
For Use With (Equipment) Automated robotic liquid handling systems
Includes C18 Coating
Quantity 10 Pk.
Supel™ is a trademark of Sigma-Aldrich Co. LLC.