Learn More
MilliporeSigma™ Supelco™ Supel™ BioSPME 96-Pin Device Positioning Adapter
Catalog No. 111023102 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Supel™ BioSPME 96-Pin Device
Catalog No. For Use With (Equipment)
11-102-3102 Supel™ BioSPME 96-Pin Device
1 options
Catalog No. 11-102-3102 Supplier MilliporeSigma™ Supelco™ Supplier No. 59686U
Add to Cart
Add to Cart
Supel™ BioSPME Positioning Adapter is used to securely and accurately position the Supel™ BioSPME 96-Pin Device.
  • The Supel™ BioSPME 96-Pin Device Positioning Adapter is used to securely and accurately position the Supel™ BioSPME 96-Pin Device into the robotic deck during the automated solid phase microextraction (SPME) workflow.
  • The adapter should be situated in a position on the automation deck where the BioSPME device will rest while not in use (i.e. before the extraction workflow begins, after the extraction workflow is completed, and/or during steps when the grippers are needed to perform an alternative action).
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Supel™ BioSPME 96-Pin Device