Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Supel™ Sphere Carbon/NH2 SPE Tube

Catalog No. 111005401 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
30/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-5401 30/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-5401 Supplier MilliporeSigma™ Supelco™ Supplier No. 54283U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SPE tube packed entirely with spherical, non-friable particles. Improved flow characteristics and faster flow for gravity filtration. Reduced susceptibility to the formation of fines

The need for SPE cartridges with improved flow characteristics and reduced susceptibility to the formation of fines has brought about the development of a family of SPE tubes packed entirely with spherical, non-friable particles. The faster, more consistent flow produced by the spherical design produces accelerated gravity filtration, thus, eliminating the need for a vacuum manifold to process samples.
TRUSTED_SUSTAINABILITY

Specifications

Column Phase Sphere Carbon/NH2
Column Type SPE
For Use With (Application) Food and Beverages
Product Line Supel
Type Solid Phase Extraction Column
Volume (Metric) 6 mL
Material Polypropylene
Quantity 30/Pk.

Supel is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.