Learn More
MilliporeSigma™ Supelco™ Supelclean™ ENVI™-18 SPE Tube, Bed Wt. 100 mg
Catalog No. 111009707 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
100 mg
Catalog No. Bed Weight
11-100-9707 100 mg
1 options
Catalog No. 11-100-9707 Supplier MilliporeSigma™ Supelco™ Supplier No. 57062
Add to Cart
Add to Cart
Supelclean ENVI-18 SPE tube is used for reversed phase separations involving a polar (usually aqueous) or moderately polar sample matrix (mobile phase) and a nonpolar stationary phase.
Retention Mechanism: Reversed-phase. Sample Matrix Compatibility: Aqueous solutions (drinking, ground, waste water). Polymerically bonded, octadecyl (17% C), endcapped. Excellent for cleaning, extracting and concentrating pollutants from aqueous environmental samples. Higher 17% C loading for increased binding capacities and higher recoveries. Higher carbon loading also offers greater resistance to extreme pH conditions. Used for extracting herbicides, fungicides, and pesticides from waste material.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase ENVI-18
Column Type SPE
Bed Weight 100 mg
For Use With (Application) Food and Beverages
Product Line Supelclean ENVI
Type Solid Phase Extraction Column
Volume (Metric) 1 mL
Endcapped Yes
Material PE Frit
Pore Size 60 Å
Quantity 108/Pk.
Surface Area (Metric) 475 m2/g
Show More Show Less
Supelclean and ENVI are trademarks of Sigma-Aldrich Co. LLC