Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Supelclean™ ENVI™-Chrom P SPE Bulk Packing

Catalog No. 111003485 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3485 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3485 Supplier MilliporeSigma™ Supelco™ Supplier No. 57217
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly crosslinked, neutral, specially cleaned styrenedivinylbenzene resin used to retain hydrophobic compounds with some hydrophilic functionality under reversed phase conditions.

Styrene/divinyl benzene co-polymer. Retention Mechanism: Reversed-phase or Adsorption. Sample Matrix Compatibility: Aqueous solutions. Particle Size: 80-160 μm; Spherical Shape; Pore Size: 110- 175 Å; Surface Area: 900 m2/g. Highly crosslinked, neutral, specially cleaned styrenedivinylbenzene resin used to retain hydrophobic compounds with some hydrophilic functionality under reversed phase conditions. Highly resistant to extreme pH conditions. Typical applications include aromatics and phenolic compounds from aqueous sample matrices. Used for priority pollutant phenols from aqueous samples.
TRUSTED_SUSTAINABILITY

Specifications

Pore Size 20 to 300 Å
Quantity 50 g
Surface Area (Metric) 900 m2/g
Product Line Supelclean
Diameter (Metric) 80 to 160 μm

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.