Learn More
MilliporeSigma™ Supelco™ Supelclean™ ENVI-Florisil™- SPE Tube, PTFE Frit, Bed Wt. 500 mg
Catalog No. 111009291 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
500 mg
Catalog No. Bed Weight
11-100-9291 500 mg
1 options
Catalog No. 11-100-9291 Supplier MilliporeSigma™ Supelco™ Supplier No. 57058
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Magnesium Silicate. Retention Mechanism: Normal phase or Adsorption. Sample Matrix Compatibility: Organic solutions. Mesh: 100/200; Available with PTFE or stainless steel frits. Tested for US Environmental Protection Agency (EPA) Contract Laboratory Program (CLP) statement of work for pesticides. Highly polar material that strongly adsorbs to polar compounds from nonpolar matrices under normal phase conditions. Typical applications include alcohols, aldehydes, amines, herbicides, pesticides, PCBs, ketones, nitro compounds, organic acids, and phenols.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase ENVI-Florisil
Column Type SPE
Bed Weight 500 mg
For Use With (Application) Food and Beverages
Product Line Supelclean ENVI Florisil
Type Solid Phase Extraction Column
Volume (Metric) 3 mL
Material PTFE Frit
Quantity 54/Pk.
Diameter (Metric) Inner 100-200 mesh
Supelclean and ENVI are trademarks of Sigma-Aldrich Co. LLC. Florisil is a registered trademark of U.S. Silica Company.