Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Supelclean™ ENVI-Florisil™- SPE Tube, Stainless Steel Frit, Bed Wt. 1 g

Catalog No. 111020801 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight
11-102-0801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0801 Supplier MilliporeSigma™ Supelco™ Supplier No. 57053
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

Magnesium Silicate. Retention Mechanism: Normal phase or Adsorption. Sample Matrix Compatibility: Organic solutions. Mesh: 100/200; Available with PTFE or stainless steel frits. Tested for US Environmental Protection Agency (EPA) Contract Laboratory Program (CLP) statement of work for pesticides. Highly polar material that strongly adsorbs to polar compounds from nonpolar matrices under normal phase conditions. Typical applications include alcohols, aldehydes, amines, herbicides, pesticides, PCBs, ketones, nitro compounds, organic acids, and phenols.
TRUSTED_SUSTAINABILITY

Specifications

Column Phase ENVI-Florisil
Column Type SPE
Bed Weight 1 g
For Use With (Application) Food and Beverages
Product Line Supelclean ENVI
Type Solid Phase Extraction Column
Volume (Metric) 6 mL
Material Stainless steel Frit
Quantity 30/Pk.
Diameter (Metric) Inner 100-200 mesh

Supelclean and ENVI are trademarks of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.