Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-18 SPE Bulk Packing
Catalog No. 111003710 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-3710 100 g
1 options
Catalog No. 11-100-3710 Supplier MilliporeSigma™ Supelco™ Supplier No. 57202
Add to Cart
Add to Cart
Endcapped
LC-18 (Octadecyl) reversed-phase Supelclean™ SPE packing may be used for the extraction of nonpolar to moderately polar analytes from aqueous samples.
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 60 Å
Quantity 100 g
Surface Area (Metric) 475 m2/g
Product Line Supelclean
Supelclean is a trademark of Sigma-Aldrich Co. LLC