Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-18 SPE Tube, Bed Wt. 100 mg
Catalog No. 111009654 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
100 mg
Catalog No. Bed Weight
11-100-9654 100 mg
1 options
Catalog No. 11-100-9654 Supplier MilliporeSigma™ Supelco™ Supplier No. 504270
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Supelclean LC-18 SPE tube is used in solid phase extraction (SPE) method to extract non-polar to moderately polar compounds from aqueous samples.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase LC-18
Column Type SPE
Bed Weight 100 mg
For Use With (Application) Food and Beverages
Product Line Supelclean
Type Solid Phase Extraction Column
Volume (Metric) 1 mL
Endcapped Yes
Material PE Frit
Pore Size 60 Å
Quantity 108/Pk.
Surface Area (Metric) 475 m2/g
Show More Show Less
Supelclean is a trademark of Sigma-Aldrich Co. LLC