Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-4 SPE Tube, Wide Pore, Bed Wt. 500 mg
Catalog No. 111008881 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
500 mg
Catalog No. Bed Weight
11-100-8881 500 mg
1 options
Catalog No. 11-100-8881 Supplier MilliporeSigma™ Supelco™ Supplier No. 57089
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Larger pore size to accommodate larger macromolecules (e.g. proteins and peptide). Commonly used for desalting and extracting proteins/peptides in aqueous samples.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase LC-4
Column Type SPE
Bed Weight 500 mg
For Use With (Application) Food and Beverages
Product Line Supelclean
Type Solid Phase Extraction Column
Volume (Metric) 3 mL
Endcapped Yes
Material PE Frit
Pore Size 500 Å
Quantity 54/Pk.
Show More Show Less
Supelclean is a trademark of Sigma-Aldrich Co. LLC