Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-8 SPE Bulk Packing
Catalog No. 111003714 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-3714 100 g
1 options
Catalog No. 11-100-3714 Supplier MilliporeSigma™ Supelco™ Supplier No. 57201
Add to Cart
Add to Cart
Endcapped
LC-8 (Octyl) reversed-phase Supelclean™ SPE packing may be used to extract nonpolar to moderately polar analytes from aqueous samples.
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 60 Å
Quantity 100 g
Surface Area (Metric) 475 m2/g
Product Line Supelclean
Supelclean is a trademark of Sigma-Aldrich Co. LLC