Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-Florisil™ SPE Bulk Packing
Catalog No. 111004437 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-4437 100 g
1 options
Catalog No. 11-100-4437 Supplier MilliporeSigma™ Supelco™ Supplier No. 57209
Add to Cart
Add to Cart
LC-Florisil™ Supelclean™™ SPE packing may be used to extract polar analytes from nonpolar samples.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 g
Product Line Supelclean
Diameter (Metric) 100 to 120 mesh
Supelclean is a trademark of Sigma-Aldrich Co. LLC