Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-NH2 SPE Bulk Packing
Catalog No. 111003669 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-3669 100 g
1 options
Catalog No. 11-100-3669 Supplier MilliporeSigma™ Supelco™ Supplier No. 57205
Add to Cart
Add to Cart
LC-NH2 (aminopropyl) normal phase Supelclean™ SPE tubes may be used to extract moderately polar to polar analytes from nonaqueous samples.
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 60 Å
Quantity 100 g
Surface Area (Metric) 475 m2/g
Product Line Supelclean
Supelclean is a trademark of Sigma-Aldrich Co. LLC