Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-Ph SPE Tube, Bed Wt. 500 mg
Catalog No. 111009852 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
500 mg
Catalog No. Bed Weight
11-100-9852 500 mg
1 options
Catalog No. 11-100-9852 Supplier MilliporeSigma™ Supelco™ Supplier No. 505269
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase LC-Ph
Column Type SPE
Bed Weight 500 mg
For Use With (Application) Food and Beverages
Product Line Supelclean
Type Solid Phase Extraction Column
Volume (Metric) 3 mL
Endcapped Yes
Material PE Frit
Pore Size 60 Å
Quantity 54/Pk.
Surface Area (Metric) 475 m2/g
Show More Show Less
Supelclean is a trademark of Sigma-Aldrich Co. LLC