Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-SCX SPE Tube, Bed Wt. 100 mg
Catalog No. 111009236 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
100 mg
Catalog No. Bed Weight
11-100-9236 100 mg
1 options
Catalog No. 11-100-9236 Supplier MilliporeSigma™ Supelco™ Supplier No. 504920
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Supelclean LC-SCX SPE Tubes consist of sulfonic acid bonded silica with Na+ counterions. They can be used for the strong cation exchange of cations, antibiotics, drugs, organic bases, amino acids, catecholamines, herbicides, nucleic acids and bases, nucleosides, and surfactants.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase LC-SCX
Column Type SPE
Bed Weight 100 mg
For Use With (Application) Food and Beverages
Product Line Supelclean
Type Solid Phase Extraction Column
Volume (Metric) 1 mL
Endcapped Yes
Material Polypropylene
Pore Size 60 Å
Quantity 108/Pk.
Diameter (Metric) Inner 40-45 μm
Show More Show Less
Supelclean is a trademark of Sigma-Aldrich Co. LLC