Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-Si Bulk Packing
Catalog No. 111004472 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-4472 100 g
1 options
Catalog No. 11-100-4472 Supplier MilliporeSigma™ Supelco™ Supplier No. 57200
Add to Cart
Add to Cart
LC-Si Adsorption SPE Tubes may be used to extract polar analytes from nonpolar samples
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 60 Å
Stationary Phase Silica Gel (Irregularly Shaped)
Quantity 100 g
Surface Area (Metric) 475 m2/g
Product Line Supelclean
Product Type Bulk Packing
For Use With (Application) Normal Phase Separation
Supelclean is a trademark of Sigma-Aldrich Co. LLC