Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-Si SPE Tube, Bed Wt. 100 mg
Catalog No. 111009790 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
100 mg
Catalog No. Bed Weight
11-100-9790 100 mg
1 options
Catalog No. 11-100-9790 Supplier MilliporeSigma™ Supelco™ Supplier No. 504041
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Supelclean LC-Si SPE Tube is used to extract polar analytes from nonpolar samples. It does not contain a bonded phase.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase LC-Si
Column Type SPE
Bed Weight 100 mg
For Use With (Application) Food and Beverages
Product Line Supelclean
Type Solid Phase Extraction Column
Volume (Metric) 1 mL
Material PE Frit
Pore Size 60 Å
Quantity 108/Pk.
Surface Area (Metric) 475 m2/g
Show More Show Less
Supelclean is a trademark of Sigma-Aldrich Co. LLC