Learn More
MilliporeSigma™ Supelco™ Supelclean™ LC-WCX SPE Tube, Bed Wt. 100 mg
Catalog No. 111009273 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
100 mg
Catalog No. Bed Weight
11-100-9273 100 mg
1 options
Catalog No. 11-100-9273 Supplier MilliporeSigma™ Supelco™ Supplier No. 505595
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Supelclean LC-WCX SPE Tube is used to adsorb moderately polar to polar compounds from nonaqueous samples.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase LC-WCX
Column Type SPE
Bed Weight 100 mg
For Use With (Application) Food and Beverages
Product Line Supelclean
Type Solid Phase Extraction Column
Volume (Metric) 1 mL
Material Polypropylene
Pore Size 60 Å
Quantity 108/Pk.
Diameter (Metric) Inner 40-45 μm
Show More Show Less
Supelclean is a trademark of Sigma-Aldrich Co. LLC