Learn More
MilliporeSigma™ Supelco™ Supelclean™ PSA SPE Bulk Packing
Catalog No. 111008741 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-8741 100 g
1 options
Catalog No. 11-100-8741 Supplier MilliporeSigma™ Supelco™ Supplier No. 52738U
Add to Cart
Add to Cart
Endcapped. Strong affinity and high capacity for removing fatty acids, organic acids, and some polar pigments and sugars when conducting multi-residue pesticide analysis in foods.
Retention Mechanism: Normal-phase and anion-exchange. Sample Matrix Compatibility: Organic or aqueous solutions. Polymerically bonded, ethylenediamine-N-propyl phase that contains both primary and secondary amines. A weak anion exchanger with a pKa of 10.1 and 10.9. Similar to aminopropyl SPE phases (NH2) in terms of selectivity, but has a much higher capacity due to presence of secondary amine (0.98-1.05 meq/g). Strong affinity and high capacity for removing fatty acids, organic acids, and some polar pigments and sugars when conducting multi-residue pesticide analysis in foods. Has been shown to significantly reduce matrix-enhancement effects encountered during the GC analysis of food products. Tested for superior cleanliness using GC-FID and GC-MS. Bidentate nature of ligand allows for chelation
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 70 Å
Quantity 100 g
Surface Area (Metric) 500 m2/g
Product Line Supelclean
Capacity (Metric) 0.98 to 1.05 meq/g capacity
Supelclean is a trademark of Sigma-Aldrich Co. LLC