Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Supelclean™ Z-Sep/C18 Mixed Bed SPE Tube , Bed Wt. 420 mg

Catalog No. 111009491 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
420 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight
11-100-9491 420 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-9491 Supplier MilliporeSigma™ Supelco™ Supplier No. 54282U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

This products was developed for pesticide residue analysis in livestock products. The Supelclean Z-Sep/C18 mixed bed tube consists of a mixed bed with Z-Sep sorbent (Zirconia coated silica gel) and ODS (C18) silica gel. Use for samples that contain high polarity pesticides.
TRUSTED_SUSTAINABILITY

Specifications

Column Phase Z-Sep/C18
Column Type SPE
Bed Weight 420 mg
For Use With (Application) Food and Beverages
Product Line Supelclean
Type Solid Phase Extraction Column
Volume (Metric) 6 mL
Material PE Frit
Quantity 30/Pk.

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.