Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Supelpak™-2 matrix styrene-divinylbenzene, 20-60 mesh

Catalog No. 111016354 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6354 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6354 Supplier MilliporeSigma™ Supelco™ Supplier No. 20279
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Purified XAD-2

Supelpak™-2 is purified Amberlite™ XAD-2™ that has been cleaned to meet and exceed US EPA-recommended criteria for purity, as outlined in Level I Environmental Assessment Procedures Manual. It is the best resin to use for standard air sampling methods requiring resin tested for background TCO (total chromatographic organics) level. Packaged in glass containers.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 20/60 Mesh
Technique Chromatography
Packaging Jar
Quantity 100 g
Surface Area (Metric) ˜300 m2/g
Product Line Supelpak
Material Styrene-divinylbenzene
Density 1.07 g/mL at 25°C (skeletal)(lit.)
CAS 9003-70-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.