Learn More
MilliporeSigma™ Supelco™ Supelpak™ -2SV matrix styrene-divinylbenzene, 20-60 mesh
Catalog No. 111005880 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 g
Catalog No. Quantity
11-100-5880 1000 g
1 options
Catalog No. 11-100-5880 Supplier MilliporeSigma™ Supelco™ Supplier No. 13674U
Add to Cart
Add to Cart
Request a Quote
Supelpak™-2SV is purified Amberlite™ XAD2 that has been specially cleaned and tested for optimal performance in the capturing and extraction of semivolatile organics. Packaged in glass containers.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 20/60 Mesh
Quantity 1000 g
Surface Area (Metric) ˜300 m2/g
Product Line Supelpak