Learn More
Sigma Aldrich Fine Chemicals Biosciences SupeltexR M-2A Capillary f
Catalog No. 502900001
Encompass
Characteristics Seals at 1/4 turn past fingertight. High reusability. Wont stick to metal or glass.Material Composition DuPont VESPEL SP-21 (85% polyimide 15% graphite)Maximum Temperature 400 C
Catalog No. 50-290-0001 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 503258
Characteristics Seals at 1/4 turn past fingertight. High reusability. Wont stick to metal or glass.Material Composition DuPont VESPEL SP-21 (85% polyimide 15% graphite)Maximum Temperature 400 C