Learn More
Sigma Aldrich Fine Chemicals Biosciences SupelTM Carbon LC 2.7 mum
Catalog No. 502909014
Encompass
Supel(TM) Carbon LC is a high-speed ultra-high performance liquid chromatography column based on 200 A… porous graphitic carbon (PGC) particle technology. The PGC particle provides a 100% carbon plane for the analytes to interact with via hydrophobic interactions and through ionic interactions via the polar retention effect on graphite (PREG) effect. This particle design exhibits high column efficiency for polar compoundsand enhanced retention of compounds typically requiring hydrophilic interaction liquid chromatography (HILIC) to retain. Through the use of patent-pending synthetic procedures a narrow particle size distribution is prevalent with this technology further enhancing the chromatographic performance that this column can display.
Catalog No. 50-290-9014 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 59984U
Supel(TM) Carbon LC is a high-speed ultra-high performance liquid chromatography column based on 200 A… porous graphitic carbon (PGC) particle technology. The PGC particle provides a 100% carbon plane for the analytes to interact with via hydrophobic interactions and through ionic interactions via the polar retention effect on graphite (PREG) effect. This particle design exhibits high column efficiency for polar compoundsand enhanced retention of compounds typically requiring hydrophilic interaction liquid chromatography (HILIC) to retain. Through the use of patent-pending synthetic procedures a narrow particle size distribution is prevalent with this technology further enhancing the chromatographic performance that this column can display.