Learn More
3M™ Super 77™ Multipurpose Spray Adhesive
Catalog No. 19070907 Shop All 3M Products
Change view
Click to view available options
Quantity:
16.75 oz.
Catalog No. Quantity
19-070-907 16.75 oz.
1 options
This item is not returnable. View return policy
Catalog No. 19-070-907 Supplier 3M™ Supplier No. 00021200212109
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
3M Company Super 77™ Multipurpose Spray Adhesive is a versatile adhesive that bonds a wide range of lightweight materials
  • Provides professional, industrial strength for demanding applications
  • Offers a fast, aggressive tack for a quick bond that reduces set time
  • Bonds paper, cardboard, fabric, insulation, plastic, metal, wood and more
  • Allows for a bond time of 15 seconds to 30 minutes
  • Repositionable while providing long-lasting strength
  • Fast-drying
  • Provides fast results to keep projects moving ahead
  • Photo-safe
  • Easy to dispense and apply directly onto surfaces and materials where needed, with no soak-in or yellowing over time
  • Net Wt 16.75 oz
  • 3M Catalog# 77
TRUSTED_SUSTAINABILITY
Specifications
Quantity 16.75 oz.