Learn More
Educational Innovations SUPER ABSORBENT POWDER 4OZ
SDP
Catalog No. NC0384948
Encompass
This fine white powder instantly turns liquid water into a slush-like solid substance. Technically speaking, the polymer absorbs from 500 to 1,000 times its own weight in water.
Catalog No. NC0384948 Supplier Educational Innovations Supplier No. GB6A
This fine white powder instantly turns liquid water into a slush-like solid substance. Technically speaking, the polymer absorbs from 500 to 1,000 times its own weight in water.