Learn More
Scytek Laboratories Super Block, 500 mL, Serum-Free Protein Block
SDP
Catalog No. NC9442419
Encompass_Preferred
Is a unique serum-free protein block for immunochemistry. Super Block eliminates the need for matching species with the link antibody and is often, more effective at reducing non-specific background staining than normal serum. This product is currently being used in a variety of applications including, Immunohistochemistry, ELISA plate blocking, and blotting. For OEM customers, this product can be modified to meet individual assay needs. Inquire about custom vialing, labeling, kit assembly and drop shipping.
Catalog No. NC9442419 Supplier Scytek Laboratories Supplier No. AAA500
Is a unique serum-free protein block for immunochemistry. Super Block eliminates the need for matching species with the link antibody and is often, more effective at reducing non-specific background staining than normal serum. This product is currently being used in a variety of applications including, Immunohistochemistry, ELISA plate blocking, and blotting. For OEM customers, this product can be modified to meet individual assay needs. Inquire about custom vialing, labeling, kit assembly and drop shipping.