Learn More
Teknova Super Broth, 1 Kg.
SDP
Catalog No. 502387969
Encompass
Super broth is a rich broth for high plasmid yield. The high levels of peptides, amino acids, vitamins and cofactors results in high yields ofEscherichia coli cells and lambda bacteriophages.
Catalog No. 50-238-7969 Supplier Teknova Supplier No. S0140
Super broth is a rich broth for high plasmid yield. The high levels of peptides, amino acids, vitamins and cofactors results in high yields ofEscherichia coli cells and lambda bacteriophages.