Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Super Broth, 1000 mL
SDP

Catalog No. NC1115169
Encompass_Preferred

Super Broth is used for the mass cultivation of Escherichia coli for high yield protein and plasmid DNA production. Autoclave sterilized Super Broth is the best commonly used E. Coli growth medium for plasmid production. Super Broth has 220% more peptone and 300% more yeast extract than LB. The major nutrients in peptone and yeast extract are still in excess well after cells reach saturation. Super Broth is is used for plasmid or protein production.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered and Autoclave Sterilized
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. NC1115169 Supplier Quality Biological Inc Supplier No. 340018131
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Super Broth is used for the mass cultivation of Escherichia coli for high yield protein and plasmid DNA production. Autoclave sterilized Super Broth is the best commonly used E. Coli growth medium for plasmid production. Super Broth has 220% more peptone and 300% more yeast extract than LB. The major nutrients in peptone and yeast extract are still in excess well after cells reach saturation. Super Broth is is used for plasmid or protein production.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered and Autoclave Sterilized
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.