Learn More
Teknova Super Broth. 1000mL, Sterile.
SDP
Catalog No. 50843031
Encompass
Super broth is a rich broth for high plasmid yield. The high levels of peptides, amino acids, vitamins and cofactors results in high yields ofEscherichia coli cells and lambda bacteriophages.
This item is not returnable. View return policy
Catalog No. 50-843-031 Supplier Teknova Supplier No. S0130
Super broth is a rich broth for high plasmid yield. The high levels of peptides, amino acids, vitamins and cofactors results in high yields ofEscherichia coli cells and lambda bacteriophages.