Learn More
Teknova Super Broth. 5 Kg
SDP
Catalog No. 502388069
Encompass
Super broth is a rich broth for high plasmid yield. The high levels of peptides, amino acids, vitamins and cofactors results in high yields ofEscherichia coli cells and lambda bacteriophages.
Catalog No. 50-238-8069 Supplier Teknova Supplier No. S0145
Super broth is a rich broth for high plasmid yield. The high levels of peptides, amino acids, vitamins and cofactors results in high yields ofEscherichia coli cells and lambda bacteriophages.