Learn More
Bioworld Super Broth, 500 g
Catalog No. 50254450
Encompass_Preferred
Super BrothRich medium for obtaining high yields of λ bacteriophage in liquid lysates Also used for higher plasmid yieldpH: 6.8-7.5Dehydrated Appearance: homogeneous powderPrepared Appearance: medium to dark, yellow to tan brothSoluble in waterContains:Soy HydrolysateYeast ExtractDipotassium PhosphateMonopotassium Phosphate
Catalog No. 50-254-450 Supplier Bioworld Supplier No. 306210261
Super BrothRich medium for obtaining high yields of λ bacteriophage in liquid lysates Also used for higher plasmid yieldpH: 6.8-7.5Dehydrated Appearance: homogeneous powderPrepared Appearance: medium to dark, yellow to tan brothSoluble in waterContains:Soy HydrolysateYeast ExtractDipotassium PhosphateMonopotassium Phosphate